accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42872-30-0

IUPAC Name: 2-(3-benzoylphenyl)propanenitrile
Synonyms: 2-(3-benzoylphenyl)propanenitrile
Molecular Weight (g/mol): 235.29
Molecular Formula: C16H13NO
InChi Key: RGYOCHMZSLUCNP-UHFFFAOYNA-N
SMILES: CC(C#N)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results