accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNNClClClClClClOCH3(E/Z)

CAS RN 42880-07-9

IUPAC Name: 2-[2-(3,4-dimethoxyphenyl)ethenyl]-4,6-bis(trichloromethyl)-1,3,5-triazine
Synonyms: 2-[2-(3,4-dimethoxyphenyl)ethenyl]-4,6-bis(trichloromethyl)-1,3,5-triazine
Molecular Weight (g/mol): 477.97
Molecular Formula: C15H11Cl6N3O2
InChi Key: ZJRNXDIVAGHETA-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C(C=CC2=NC(=NC(=N2)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results