accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42882-31-5

IUPAC Name: 1-(naphthalen-1-yl)ethan-1-amine
Synonyms: 1-(naphthalen-1-yl)ethanamine
Molecular Weight (g/mol): 171.24
Molecular Formula: C12H13N
InChi Key: RTCUCQWIICFPOD-UHFFFAOYNA-N
SMILES: CC(N)C1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)