accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 429-06-1

IUPAC Name: tetraethylazanium; tetrafluoroboranuide
Synonyms: tetraethylammonium tetrafluoroborate
Molecular Weight (g/mol): 217.06
Molecular Formula: C8H20BF4N
InChi Key: XJRAKUDXACGCHA-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CC[N+](CC)(CC)CC
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (7)
  • (2)
  • (8)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)