accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSClF

CAS RN 42908-73-6

IUPAC Name: 4-fluorophenyl chloromethanethioate
Synonyms: 4-fluorophenyl chloromethanethioate
Molecular Weight (g/mol): 190.62
Molecular Formula: C7H4ClFOS
InChi Key: HZBXSUFBKXPOAF-UHFFFAOYSA-N
SMILES: FC1=CC=C(OC(Cl)=S)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results