accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42908-86-1

IUPAC Name: 2-(chloromethyl)benzoyl chloride
Synonyms: 2-(chloromethyl)benzoyl chloride
Molecular Weight (g/mol): 189.04
Molecular Formula: C8H6Cl2O
InChi Key: TXZFBHYDQGYOIT-UHFFFAOYSA-N
SMILES: ClCC1=CC=CC=C1C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results