accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CI

CAS RN 4292-19-7

IUPAC Name: 1-iodododecane
Synonyms: 1-iodododecane
Molecular Weight (g/mol): 296.24
Molecular Formula: C12H25I
InChi Key: GCDPERPXPREHJF-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCI
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (1)
  • (8)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (1)
  • (6)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (7)
  • (1)
  • (8)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)