accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 4292-75-5

IUPAC Name: hexylcyclohexane
Synonyms: 1-cyclohexylhexane
Molecular Weight (g/mol): 168.32
Molecular Formula: C12H24
InChi Key: QHWAQXOSHHKCFK-UHFFFAOYSA-N
SMILES: CCCCCCC1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results