accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4294-57-9

IUPAC Name: bromo(4-methylphenyl)magnesium
Synonyms: bromo(4-methylphenyl)magnesium
Molecular Weight (g/mol): 195.34
Molecular Formula: C7H7BrMg
InChi Key: ZRJNGFJIBZKXTP-UHFFFAOYSA-M
SMILES: CC1=CC=C([Mg]Br)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)