accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42967-55-5

IUPAC Name: potassium 4-(methoxycarbonyl)benzoate
Synonyms: potassium 4-(methoxycarbonyl)benzoate
Molecular Weight (g/mol): 218.25
Molecular Formula: C9H7KO4
InChi Key: DWNDFZTVSFSFTB-UHFFFAOYSA-M
SMILES: [K+].COC(=O)C1=CC=C(C=C1)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results