accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4298-52-6

CHS

CAS RN 4298-52-6

IUPAC Name: 2-ethynylthiophene
Synonyms: 2-ethynylthiophene
Molecular Weight (g/mol): 108.16
Molecular Formula: C6H4S
InChi Key: LWISLHRIEATKTM-UHFFFAOYSA-N
SMILES: C#CC1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)