accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42989-85-5

IUPAC Name: 2-({[(tert-butoxy)carbonyl]amino}oxy)acetic acid
Synonyms: {[(tert-butoxycarbonyl)amino]oxy}acetic acid
Molecular Weight (g/mol): 191.18
Molecular Formula: C7H13NO5
InChi Key: QBXODCKYUZNZCY-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NOCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (4)
  • (2)
Quantity 
  • (4)
  • (1)
  • (4)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (4)
  • (2)
  • (4)
  • (1)
  • (4)
  • (2)