accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OF

CAS RN 430-51-3

IUPAC Name: 1-fluoropropan-2-one
Synonyms: fluoroacetone
Molecular Weight (g/mol): 76.07
Molecular Formula: C3H5FO
InChi Key: MSWVMWGCNZQPIA-UHFFFAOYSA-N
SMILES: CC(=O)CF
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)