accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NFF

CAS RN 430-67-1

IUPAC Name: 2,2-difluoroethan-1-amine
Synonyms: 2,2-difluoroethanamine
Molecular Weight (g/mol): 81.07
Molecular Formula: C2H5F2N
InChi Key: OVRWUZYZECPJOB-UHFFFAOYSA-N
SMILES: NCC(F)F
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)