CAS RN 43021-26-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ Anhydroecgonine Methyl Ester Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Suitable for use in LC/MS or GC/MS applications for urine drug testing, clinical toxicology, or forensic analysis.