accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHNNH2

CAS RN 43024-15-3

IUPAC Name: 4-ethyl-1H-pyrazol-5-amine
Synonyms: 4-ethyl-2H-pyrazol-3-amine
Molecular Weight (g/mol): 111.15
Molecular Formula: C5H9N3
InChi Key: RDCODVKTTJWFAR-UHFFFAOYSA-N
SMILES: CCC1=C(N)NN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results