accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4303-67-7

IUPAC Name: 1-dodecyl-1H-imidazole
Synonyms: N-laurylimidazole
Molecular Weight (g/mol): 236.4
Molecular Formula: C15H28N2
InChi Key: JMTFLSQHQSFNTE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCN1C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results