accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COCl

CAS RN 43076-61-5

IUPAC Name: 1-(4-tert-butylphenyl)-4-chlorobutan-1-one
Synonyms: 1-(4-tert-butylphenyl)-4-chlorobutan-1-one
Molecular Weight (g/mol): 238.76
Molecular Formula: C14H19ClO
InChi Key: RLKSQLJFGCDUOX-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C(=O)CCCCl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results