accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH2

CAS RN 4312-99-6

IUPAC Name: oct-1-en-3-one
Synonyms: oct-1-en-3-one
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: KLTVSWGXIAYTHO-UHFFFAOYSA-N
SMILES: CCCCCC(=O)C=C
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)