accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOClNNN

CAS RN 43121-43-3

IUPAC Name: 1-(4-chlorophenoxy)-3,3-dimethyl-1-(1H-1,2,4-triazol-1-yl)butan-2-one
Synonyms: typhon adifon bayleton triple triadimefon mighty nurex otria 25 strike bayleton BM bayleton CF
Molecular Weight (g/mol): 293.75
Molecular Formula: C14H16ClN3O2
InChi Key: WURBVZBTWMNKQT-UHFFFAOYNA-N
SMILES: CC(C)(C)C(=O)C(OC1=CC=C(Cl)C=C1)N1C=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results