accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFCl

CAS RN 43141-66-8

IUPAC Name: 1-chloro-4-(difluoromethyl)benzene
Synonyms: 1-chloro-4-(difluoromethyl)benzene
Molecular Weight (g/mol): 162.56
Molecular Formula: C7H5ClF2
InChi Key: UNSMMYPOWHMIRS-UHFFFAOYSA-N
SMILES: FC(F)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results