accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4316-57-8

IUPAC Name: 4-nitro-N,N-diphenylaniline
Synonyms: 4-nitro-N,N-diphenylaniline
Molecular Weight (g/mol): 290.32
Molecular Formula: C18H14N2O2
InChi Key: UQOKZDUUBVGFAK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results