accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4316-65-8

IUPAC Name: 3,5,5-trimethylhex-1-ene
Synonyms: 3,5,5-trimethylhex-1-ene
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: JTXUVHFRSRTSAT-UHFFFAOYNA-N
SMILES: CC(CC(C)(C)C)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results