accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 43192-31-0

IUPAC Name: 2-bromo-4-methoxybenzaldehyde
Synonyms: 2-bromo-4-methoxybenzaldehyde
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: ODISAUHBLBVQKC-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=C(C=O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results