accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH3CCH3

CAS RN 432-25-7

IUPAC Name: 2,6,6-trimethylcyclohex-1-ene-1-carbaldehyde
Synonyms: β-cyclocitral
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: MOQGCGNUWBPGTQ-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C(C)(C)CCC1
Percent Purity 
  • (2)
  • (1)
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)