accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSOOOCH3NNH3C

CAS RN 43222-48-6

IUPAC Name: 1,2-dimethyl-3,5-diphenyl-1H-pyrazol-2-ium methyl sulfate
Synonyms: difenzoquat methyl sulfate(1-)
Molecular Weight (g/mol): 360.43
Molecular Formula: C18H20N2O4S
InChi Key: XQEMNBNCQVQXMO-UHFFFAOYSA-M
SMILES: COS([O-])(=O)=O.CN1C(=CC(C2=CC=CC=C2)=[N+]1C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results