accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrH3CNCH3H3CCH3

CAS RN 4328-13-6

IUPAC Name: tetrahexylazanium bromide
Synonyms: tetrahexylazanium bromide
Molecular Weight (g/mol): 434.59
Molecular Formula: C24H52BrN
InChi Key: SYZCZDCAEVUSPM-UHFFFAOYSA-M
SMILES: [Br-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (5)
  • (2)
  • (6)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)