accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFF

CAS RN 433-95-4

IUPAC Name: 1,2-bis(trifluoromethyl)benzene
Synonyms: 1,2-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 214.11
Molecular Formula: C8H4F6
InChi Key: XXZOEDQFGXTEAD-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results