accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 433316-37-1

IUPAC Name: N-(2-chloro-4-iodophenyl)-2-fluorobenzamide
Synonyms: N-(2-chloro-4-iodophenyl)-2-fluorobenzamide
Molecular Weight (g/mol): 375.57
Molecular Formula: C13H8ClFINO
InChi Key: HMFPZLBLOKLAPU-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C(=O)NC1=C(Cl)C=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results