accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4335-90-4

IUPAC Name: 3-(phenylmethylidene)pentane-2,4-dione
Synonyms: benzal acetylacetone
Molecular Weight (g/mol): 188.23
Molecular Formula: C12H12O2
InChi Key: NYRGMNMVISROGJ-UHFFFAOYSA-N
SMILES: CC(=O)C(=CC1=CC=CC=C1)C(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results