accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFO

CAS RN 434-45-7

IUPAC Name: 2,2,2-trifluoro-1-phenylethan-1-one
Synonyms: trifluoroacetophenone
Molecular Weight (g/mol): 174.12
Molecular Formula: C8H5F3O
InChi Key: KZJRKRQSDZGHEC-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (6)
Quantity 
  • (3)
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (2)
  • (6)
  • (3)
  • (3)
  • (3)
  • (2)
  • (6)
  • (5)
  • (2)
  • (3)
  • (6)
  • (3)
  • (4)
  • (4)
  • (5)