accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 434-84-4

IUPAC Name: 9H,9'H,10H,10'H-[9,9'-bianthracene]-10,10'-dione
Synonyms: 9H,9'H-[9,9'-bianthracene]-10,10'-dione
Molecular Weight (g/mol): 386.45
Molecular Formula: C28H18O2
InChi Key: ZQXZUOJNJXNUEO-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C(C2C3=CC=CC=C3C(=O)C3=CC=CC=C23)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results