accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 434-85-5

IUPAC Name: 10H,10'H-[9,9'-bianthracenylidene]-10,10'-dione
Synonyms: [9,9'-bianthracenylidene]-10,10'-dione
Molecular Weight (g/mol): 384.43
Molecular Formula: C28H16O2
InChi Key: MGRRGKWPEVFJSH-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C(C2=CC=CC=C12)=C1C2=CC=CC=C2C(=O)C2=CC=CC=C12
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)