accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3C

CAS RN 4341-76-8

IUPAC Name: ethyl but-2-ynoate
Synonyms: but-2-ynoic acid ethyl ester
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: FCJJZKCJURDYNF-UHFFFAOYSA-N
SMILES: CCOC(=O)C#CC
Boiling Point 
  • (2)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)