accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3ClSiCH3CH3Cl

CAS RN 4342-61-4

IUPAC Name: 1,2-dichloro-1,1,2,2-tetramethyldisilane
Synonyms: 1,2-dichloro-1,1,2,2-tetramethyldisilane
Molecular Weight (g/mol): 187.21
Molecular Formula: C4H12Cl2Si2
InChi Key: SFAZXBAPWCPIER-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)[Si](C)(C)Cl
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)