accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH2

CAS RN 4344-55-2

IUPAC Name: 4-butoxyaniline
Synonyms: benzenamine, 4-butoxy-
Molecular Weight (g/mol): 165.24
Molecular Formula: C10H15NO
InChi Key: UBRIHZOFEJHMIT-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(N)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)