accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4347-31-3

IUPAC Name: (2-formylthiophen-3-yl)boronic acid
Synonyms: 2-formylthiophen-3-ylboronic acid
Molecular Weight (g/mol): 155.96
Molecular Formula: C5H5BO3S
InChi Key: BBENFHSYKBYWJX-UHFFFAOYSA-N
SMILES: OB(O)C1=C(SC=C1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results