accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 434935-69-0

IUPAC Name: 2-methyl-6-nitrobenzoyl 2-methyl-6-nitrobenzoate
Synonyms: 2-methyl-6-nitrobenzoyl 2-methyl-6-nitrobenzoate
Molecular Weight (g/mol): 344.28
Molecular Formula: C16H12N2O7
InChi Key: YEKPNMQQSPHKBP-UHFFFAOYSA-N
SMILES: CC1=C(C(=O)OC(=O)C2=C(C)C=CC=C2[N+]([O-])=O)C(=CC=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results