accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 434937-12-9

IUPAC Name: 1-(cyanomethyl)piperidin-1-ium; tetrafluoroboranuide
Synonyms: 1-(cyanomethyl)piperidin-1-ium tetrafluoroborate
Molecular Weight (g/mol): 212
Molecular Formula: C7H13BF4N2
InChi Key: HSIKBRNFKUNHIN-UHFFFAOYSA-O
SMILES: F[B-](F)(F)F.N#CC[NH+]1CCCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results