CAS RN 4350-84-9
IUPAC Name:
2-oxabicyclo[2.2.2]octan-3-one
Synonyms:
2-oxabicyclo[2.2.2]octan-3-one
Molecular Weight (g/mol):
126.16
Molecular Formula:
C7H10O2
InChi Key:
NVORRKXXHHFWDH-UHFFFAOYSA-N
SMILES:
O=C1OC2CCC1CC2
1
–
1
of
1
results
1
4-Hydroxy-1-cyclohexanecarboxylic Acid delta-Lactone 98.0+%, TCI America™
CAS: 4350-84-9 Molecular Formula: C7H10O2 Molecular Weight (g/mol): 126.155 MDL Number: MFCD06797111 InChI Key: NVORRKXXHHFWDH-UHFFFAOYSA-N Synonym: 3,6-Ethanotetrahydropyran-2-one, 2-Oxabicyclo[2.2.2]octan-3-one PubChem CID: 138195 IUPAC Name: 3-oxabicyclo[2.2.2]octan-2-one SMILES: C1CC2CCC1C(=O)O2