accessibility menu, dialog, popup

UserName

Viewing profile as user:
BH3CCH3CH3CH3CH3H3CF

CAS RN 436-59-9

IUPAC Name: fluorobis(2,4,6-trimethylphenyl)borane
Synonyms: fluorobis(2,4,6-trimethylphenyl)borane
Molecular Weight (g/mol): 268.18
Molecular Formula: C18H22BF
InChi Key: WZWGERGANZMXOM-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(B(F)C2=C(C)C=C(C)C=C2C)C(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results