accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 437-17-2

IUPAC Name: hexafluoro-λ⁵-phosphanuide; triphenylmethylium
Synonyms: triphenylmethyl hexafluorophosphate
Molecular Weight (g/mol): 388.29
Molecular Formula: C19H15F6P
InChi Key: IBTFOFOFRZKIJU-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)