accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 438056-69-0

IUPAC Name: 4-(4-aminophenyl)morpholin-3-one
Synonyms: 4-(4-aminophenyl)morpholin-3-one
Molecular Weight (g/mol): 192.22
Molecular Formula: C10H12N2O2
InChi Key: MHCRLDZZHOVFEE-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)N1CCOCC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results