CAS RN 4383-08-8
IUPAC Name:
2-methyl-1-phenylprop-2-en-1-ol
Synonyms:
2-methyl-1-phenylprop-2-en-1-ol
Molecular Weight (g/mol):
148.21
Molecular Formula:
C10H12O
InChi Key:
ZGYBYYJGIKPBFD-UHFFFAOYNA-N
SMILES:
CC(=C)C(O)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
2-Methyl-1-phenyl-2-propen-1-ol, tech. 85%
2-Methyl-1-phenyl-2-propen-1-ol, tech. 85%
CAS: 4383-08-8 Molecular Formula: C10H12O Molecular Weight (g/mol): 148.21 MDL Number: MFCD00046630 InChI Key: ZGYBYYJGIKPBFD-UHFFFAOYNA-N Synonym: 2-methyl-1-phenyl-2-propen-1-ol,acmc-1bn9z,alpha-isopropenylbenzenemethanol,1-phenyl-2-methyl-2-propen-1-ol,2,6-dichloro-9-methyl-10h-purine PubChem CID: 138211 IUPAC Name: 2-methyl-1-phenylprop-2-en-1-ol SMILES: CC(=C)C(C1=CC=CC=C1)O