accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHO

CAS RN 4385-76-6

IUPAC Name: 4-(pyridin-4-yl)benzoic acid
Synonyms: 4-(pyridin-4-yl)benzoic acid
Molecular Weight (g/mol): 199.21
Molecular Formula: C12H9NO2
InChi Key: DZLGZIGLHCRIMF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=CC=NC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results