accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFN

CAS RN 439-88-3

IUPAC Name: 5-fluoro-8-methoxyquinoline
Synonyms: 5-fluoro-8-methoxyquinoline
Molecular Weight (g/mol): 177.18
Molecular Formula: C10H8FNO
InChi Key: XNOHPRHYFXAKPA-UHFFFAOYSA-N
SMILES: COC1=C2N=CC=CC2=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results