accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 439081-52-4

IUPAC Name: methyl[(oxan-4-yl)methyl]amine
Synonyms: methyl(oxan-4-ylmethyl)amine
Molecular Weight (g/mol): 129.2
Molecular Formula: C7H15NO
InChi Key: WMBCUXKYKVTJRF-UHFFFAOYSA-N
SMILES: CNCC1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results