accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4394-00-7

IUPAC Name: 2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid
Synonyms: niflumic acid upsa conseil brand of niflumic acid acidum niflumicum acido niflumico niflam flogovital upsa brand 2 of niflumic acid NFA NFL nifluminic acid
Molecular Weight (g/mol): 282.22
Molecular Formula: C13H9F3N2O2
InChi Key: JZFPYUNJRRFVQU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CN=C1NC1=CC=CC(=C1)C(F)(F)F
Percent Purity 
  • (1)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)
Product Line 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)