accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONO

CAS RN 4394-85-8

IUPAC Name: morpholine-4-carbaldehyde
Synonyms: formyl morpholine
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: LCEDQNDDFOCWGG-UHFFFAOYSA-N
SMILES: O=CN1CCOCC1
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (2)
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (2)
  • (3)
  • (3)
  • (5)
  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)