accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 439797-69-0

IUPAC Name: 4,4'-dibromo-2-nitro-1,1'-biphenyl
Synonyms: 4,4'-dibromo-2-nitro-1,1'-biphenyl
Molecular Weight (g/mol): 357
Molecular Formula: C12H7Br2NO2
InChi Key: FMSJGXRUJCWSJL-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=CC(Br)=C1)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results